Learn More
Cayman Chemical OlIgomycIn A 10mg
SDP
Supplier:  Cayman Chemical 1134210 MG
Encompass_Preferred
An inhibitor of mitochondrial F1FO ATP synthase that induces apoptosis in a variety of cell types (average GI50 = 270 nM); exhibits antifungal, antitumor, and nematocidal activities, but has poor solubility in water and other biocompatible solvents
Catalog No. 50-295-2969