Learn More
Medchemexpress LLC Oligomycin A | 579-13-5 | 99.91% | 791.06 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16589100MG
Encompass_Preferred
Oligomycin A (MCH 32), produced by Streptomyces, functions as a mitochondrial F0F1-ATPase inhibitor with a Ki of 1 μM. It also demonstrates anti-fungal activity. For research use only; not sold to patients.
  • Acts as a mitochondrial F0F1-ATPase inhibitor (Ki: 1 μM).
  • Exhibits anti-fungal activity.
  • Shows cytotoxic effects on NCI-60 cell lines with a GI50 of 10 nM.
  • Inhibits the activity of Triton X-100-solubilized ATPase with a Ki of 0.1 μM.
  • Inhibits the ability of spermatozoa to achieve feasible in vitro capacitation (IVC) at 2.4 μM.
  • Suppresses progesterone-induced in vitro acrosome exocytosis (IVAE) and associated O2 consumption and ATP levels.
Catalog No. 50-003-93474