Learn More
Selleck Chemical LLC Oligomycin A 5mg 579-13-5 MCH 32
SDP
Supplier:  Selleck Chemical LLC S14785MG
Encompass_Preferred
Oligomycin A (MCH 32) is an inhibitor of ATP synthase, inhibits oxidative phosphorylation and all the ATP-dependent processes occurring on the coupling membrane of mitochondria. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1302