Learn More
Enzo Life Sciences Oligomycin, (5mg), CAS Number: 1404-19-9
SDP
Supplier:  Enzo Life Sciences ALX380037M005
Encompass_Preferred
F-ATPase inhibitor. Macrolide antibiotic that inhibits membrane bound mitochondrial ATPase (F1), preventing phosphoryl group transfer. Induces apoptosis. MW: 791.1 (Oligomycin A). CAS Number: 1404-19-9. MI: 14: 6833. Purity: ≥90% (HPLC, TLC). Appearance: White to faint yellow powder. Solubility: Soluble in 100% ethanol, methanol (10mg/ml) or acetone (50mg/ml). Long Term Storage: +4°C
Catalog No. 50-103-1746