Learn More
Medchemexpress LLC Olinciguat | 1628732-62-6 | 98.4% | 509.39 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10906650MG
Encompass_Preferred
Olinciguat (IW-1701) is an oral guanylate cyclase (sGC) stimulator that shows concentration-dependent stimulation of sGC in purified rat and human enzyme assays and a whole cell assay. This product is for research use only and not sold to patients.
  • Oral guanylate cyclase (sGC) stimulator
  • Stimulates cGMP synthesis
Catalog No. 50-003-43316