Learn More
Medchemexpress LLC Olinciguat | 1628732-62-6 | 98.4% | 509.39 | 50 MG

Supplier: Medchemexpress LLC HY109066100MG
Olinciguat (IW-1701) is an oral guanylate cyclase (sGC) stimulator that shows concentration-dependent stimulation of sGC in purified rat and human enzyme assays, as well as in a whole cell assay. This compound is provided as a solid, typically white to light yellow in appearance.
- Oral guanylate cyclase (sGC) stimulator
- Demonstrates concentration-dependent sGC stimulation
- Suitable for in vitro enzyme and whole cell assays
- Available as a solid, typically white to light yellow
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- In vitro solubility in DMSO: 125 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.