Learn More
Selleck Chemical LLC Olitigaltin S0471-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S047110MM/1ML
Encompass_Preferred
Olitigaltin (TD-139 GB0-139) is a potent and selective inhibitor of galectin-3 with Kd of 0 036 M over galectin-1 and galectin-7 with Kd of 2 2 M and 32 M respectively TD-139 is used potentially for the treatment of idiopathic pulmonary fibrosis (IPF)
Catalog No. 50-000-43226