Learn More
Apexbio Technology LLC Olivetol 500-66-3 100mg
SDP
Supplier:  Apexbio Technology LLC C7059100
Encompass_Preferred
Olivetol (CAS 500-66-3) is a naturally occurring polyphenolic compound that acts as a competitive inhibitor of cannabinoid receptors CB1 and CB2 In vitro studies demonstrate its inhibitory effects on cytochrome P450 enzymes CYP2C19 and CYP2D6 with IC50 values of 15 3 M and 7 21 M and Ki values of 2 71 M and 2 87 M respectively Olivetol is utilized in research investigating cannabinoid-mediated signaling pathways and the modulation of metabolic enzymes involved in xenobiotic biotransformation
Catalog No. 50-000-94407