Learn More
Medchemexpress LLC Olivetolic acid | 491-72-5 | 99.9% | 224.25 g/mol | C12H16O4 | 1 ML

Supplier: Medchemexpress LLC HYW09029210MM/1ML
Olivetolic acid is a small-molecule research reagent used as a biosynthetic precursor in cannabinoid-related studies. It is supplied as a high-purity solid and as a ready-to-use 10 mM solution in DMSO for biochemical and pharmacological research.
- Biosynthetic precursor of cannabinol acid (CBGA)
- Available as solid and as 10 mM 1 mL solution in DMSO
- High purity: 99.9%
- Molecular formula: C12H16O4; molecular weight: 224.25 g/mol
- Physical form: white to off-white solid
- Documentation available: safety data sheet and certificate of analysis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.