Learn More
Medchemexpress LLC Olivetolic acid | 491-72-5 | MFCD28383644 | 99.9% | 224.25 g/mol | C12H16O4 | 5 MG

Supplier: Medchemexpress LLC HYW0902925MG
Olivetolic acid is a biosynthetic precursor of cannabinol acid (CBGA) and has demonstrated modest anticonvulsant activity in a mouse model of Dravet syndrome. It is supplied as a high-purity research reagent for biochemical and pharmacological studies.
- Biosynthetic precursor to cannabinoid acid (CBGA).
- Reported modest anticonvulsant activity in a mouse model.
- High purity (99.9%).
- Molecular formula C12H16O4 and molecular weight 224.25 g/mol.
- Available as a 5 mg solid and as a 10 mM, 1 mL solution in DMSO.
- Intended for research use in biochemical and pharmacological applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.