Learn More
Medchemexpress LLC Olmesartan methyl ester | 1347262-29-6 | MFCD24386461 | 95.0% | 460.53 g/mol | C25H28N6O3 | 5 MG

Supplier: Medchemexpress LLC HY1312785MG
Olmesartan methyl ester is a chemical intermediate used in the synthesis of olmesartan medoxomil, an angiotensin AT1 receptor antagonist. It is intended for research and synthetic applications and is supplied in small laboratory pack sizes.
- Intermediate in the synthesis of olmesartan medoxomil.
- CAS number 1347262-29-6 for unambiguous identification.
- Molecular formula C25H28N6O3 and molecular weight 460.53 g/mol.
- Supplied with stated purity of 95.0%.
- Available in small research pack sizes (5 mg).
- Typically accompanied by a certificate of analysis and analytical data for batch traceability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.