Learn More
Medchemexpress LLC HY-131278 1mg Medchemexpress, Olmesartan methyl ester CAS:1347262-29-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1312781MG
Encompass_Preferred
Medchemexpress, HY-131278 1mg Olmesartan methyl ester CAS:1347262-29-6 Olmesartan methyl ester is an intermediate in the synthesis of Olmesartan medoxomil. Olmesartan medoxomil is a potent and selective angiotensin AT1 receptor antagonist with IC50 of 66.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1270