Learn More
Selleck Chemical LLC Olmutinib
SDP
Supplier:  Selleck Chemical LLC S82945MG
Encompass_Preferred
Olmutinib (BI 1482694) is a novel third-generation epidermal growth factor receptor (EGFR) mutation-specific tyrosine kinase inhibitor (TKI) Also a potent inhibitor of Bruton s tyrosine kinase
Catalog No. 50-000-00125