Learn More
Medchemexpress LLC Olodaterol | 868049-49-4 | 99.9% | C21H26N2O5 | 2 MG

Supplier: Medchemexpress LLC HY143012MG
Olodaterol, also known as BI1744, is a chemical compound with a molecular formula of C21H26N2O5 and a molecular weight of 386.44. It is supplied as an off-white to yellow solid with a purity of 99.9%.
- High purity of 99.9%
- Soluble in water at 100 mg/mL, requiring ultrasonic and warming for dissolution
- Soluble in DMSO at 100 mg/mL, requiring ultrasonic
- Optimal storage for powder is -20°C for up to 3 years or 4°C for up to 2 years
- Optimal storage for solutions is -80°C for up to 2 years or -20°C for up to 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.