Learn More
Medchemexpress LLC Olodaterol (BI1744) | 868049-49-4 | 99.9% | C21H26N2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1430150MG
Encompass_Preferred
Olodaterol (BI1744) is a selective, long-acting β2-adrenoceptor (β2-AR) agonist with an EC50 of 0.1 nM and a pKi of 9.14 for human β2-adrenoceptor. It is used for chronic obstructive pulmonary disease (COPD) and pulmonary fibrosis. Olodaterol exhibits subnanomolar affinity for the β2-AR and is selective compared to β1-AR and β3-AR subtypes.
  • Attenuates growth factor-induced motility and proliferation in fibroblasts.
  • Interferes with FGF-induced phosphorylation of signaling cascades.
  • Increases intracellular cAMP in a concentration-dependent manner.
  • Inhibits PICP increase with maximal efficacy of 70% at 10 nM.
  • Accelerates body weight recovery and attenuates TGF-β-induced lung fibrosis in C57BL/6 mice.
  • Induces a dose-dependent bronchoprotection in guinea pigs and dogs.
Catalog No. 50-003-56430