Learn More
Selleck Chemical LLC Olodaterol hydrochloride
SDP
Supplier:  Selleck Chemical LLC S592525MG
Encompass_Preferred
Olodaterol hydrochloride (BI-1744) is the hydrochloride salt form of Olodaterol which is a long-acting beta2-adrenergic agonist
Catalog No. 50-000-03942