Learn More
TARGETMOL CHEMICALS INC OLODATEROL HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T51595MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Olodaterol hydrochloride (BI-1744 HCl) is a novel long-acting (beta)2-adrenergic agonist (EC50s 97.7 nM for the human (beta)2-adrenoceptor) that exerts its pharmacological effect by binding and activating (beta)2-adrenergic receptors. purity: 99%
Catalog No. 50-257-6704