Learn More
Medchemexpress LLC Olodaterol hydrochloride | 869477-96-3 | 99.4% | C₂₁H₂₇ClN₂O₅ | 100 MG

Supplier: Medchemexpress LLC HY14301AR100MG
Olodaterol hydrochloride is a selective, long-acting β2-adrenoceptor agonist, primarily utilized in research for chronic obstructive pulmonary disease (COPD) and pulmonary fibrosis studies. This chemical is intended for laboratory use.
- White to off-white solid appearance.
- Purity of 99.36% confirmed by HPLC.
- Stable under recommended storage conditions.
- Store at 4°C, sealed and dry, away from light and moisture.
- Identified for laboratory chemical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.