Learn More
TARGETMOL CHEMICALS INC OLOROFIM 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T273005MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Olorofim is a novel selective antifungal compound with inhibitory effect on Aspergillus fumigatus DHODH (IC50 44 nM) and almost no inhibitory effect on human DHODH (>100 uM).Olorofim has good efficacy against various pathogenic filamentous and dimorphic fungi such as Penicillium spp Dermatophilus spp and Fusarium spp. purity: 98%
Catalog No. 50-258-0630