Learn More
Medchemexpress LLC Olpasiran sodium | 2225856-03-9 | C490H610F11N164Na42O306P41S7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY153492A1MG
Encompass_Preferred
Olpasiran sodium is an N-acetyl galactosamine (GalNAc)-conjugated, hepatocyte-targeted siRNA. It directly inhibits LPA messenger RNA translation in hepatocytes and potently reduces Lp(a) concentration. It can be used for the research of cardiovascular disease, such as atherosclerosis.
  • N-acetyl galactosamine (GalNAc)-conjugated, hepatocyte-targeted small interfering RNA.
  • Potently reduces lipoprotein(a) concentration by directly inhibiting LPA messenger RNA translation.
  • Demonstrated efficacy in silencing LPA reporter gene expression in human hepatocellular carcinoma cells.
  • Achieves sustained and significant reductions in serum lipoprotein(a) levels in preclinical animal models.
  • Potential application in cardiovascular disease research, specifically atherosclerosis.
Catalog No. 50-003-72823