Learn More
Cayman Chemical OlsalazIn-13C6 1mg
SDP
Supplier:  Cayman Chemical 337981 MG
Encompass_Preferred
An internal standard for the quantification of olsalazine by GC- or LC-MS
Catalog No. 50-295-0091