Learn More
Fine Science Tools Olsen-Hegar Needle Holders with Suture Cutters, Tungsten Carbide, 14 cm
SDP
Supplier:  Fine Science Tools 1250214
Encompass
Product: Olsen-Hegar Needle Holder with Scissors/ Tip Shape: Straight/ Tip Width: 2mm/ Clamping Length: 9.5mm/ Scissors: Yes/ Lock: Yes/ Alloy: Stainless Steel/ Length: 14cm/ Serrated: Yes/ Feature: Tungsten Carbide/ Unit: Each
Catalog No. NC9829741