Learn More
Apexbio Technology LLC Oltipraz 64224-21-1 200mg
SDP
Supplier:  Apexbio Technology LLC B5958200
Encompass_Preferred
Oltipraz (CAS 64224-21-1) is a small-molecule activator targeting the nuclear factor erythroid 2 related factor 2 (Nrf2) signaling pathway It is designed to induce phase II detoxifying enzymes thereby modulating cellular defense mechanisms against xenobiotic and carcinogen exposure Oltipraz exerts its biological activity primarily through upregulation of phase II enzymes including glutathione S-transferase (GST) and NAD(P)H quinone oxidoreductase (NQO1) In rat hepatocyte assays Oltipraz demonstrates enzyme induction activity with an IC50 of approximately 10 30 M Based on these pharmacological properties Oltipraz holds research potential in chemoprevention and protection against carcinogen-related toxicity by modulation of Nrf2 signaling pathways
Catalog No. 50-000-95410