Learn More
Medchemexpress LLC Oltipraz | 64224-21-1 | 99.9% | 226.34 | C8H6N2S3 | 200MG

Supplier: Medchemexpress LLC HY12519200MG
Oltipraz is a research-grade small molecule that functions as an Nrf2 activator and HIF-1α inhibitor. Supplied as a brown-to-red solid powder for laboratory research, it is intended for use in mechanistic studies of oxidative stress, phase II detoxification, and hypoxia signaling; not for human consumption.
- High purity (99.9%).
- Molecular formula C8H6N2S3; molecular weight 226.34 g/mol.
- Available in multiple pack sizes including 200 mg vials.
- Solid, brown-to-red appearance suitable for standard laboratory handling.
- Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (1 year) or -20°C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.