Learn More
Medchemexpress LLC HY-114226 10mg Medchemexpress, Olutasidenib CAS:1887014-12-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11422610MG
Encompass_Preferred
Medchemexpress, HY-114226 10mg Olutasidenib CAS:1887014-12-1 Olutasidenib is a highly potent, selective inhibitor of mutant Isocitrate dehydrogenase 1 (IDH1) that could be used in the treatment of acute myeloid leukemia (AML) or myelodysplastic syndrome (MDS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2472