Learn More
TARGETMOL CHEMICALS INC OLVEREMBATINIB 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T307125MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Olverembatinib (GZD 824) is an orally bioavailable Bcr-Abl inhibitor for Bcr-Abl(WT IC50 0.34 nM) and Bcr-Abl(T315I IC50 0.68 nM). purity: 99%
Catalog No. 50-258-0394