Learn More
AdipoGen OM173-alphaA (5 mg)
SDP
Supplier:  AdipoGen AGCN20158M005
Encompass_Preferred
Chemical. CAS: 58286-56-9. Formula: C17H16O6. MW: 316.3. Isolated from Streptomyces sp. Antibiotic. Antimycoplasma. Antifungal compound. Anti-malarial agent. Ras-competitive non-CAAX mimetic type farnesyltransferase (FTase) inhibitor. Potential anticancer compound. Inducer of antiproliferative effects in tumor cell lines. Selective DNA (cytosine-5)-methyltransferase 3B (DNMT3B) inhibitor.
Catalog No. 50-205-3545