Learn More
Medchemexpress LLC Omadacycline-d9 | 2272886-41-4 | 99.21% | 565.71 | 100 UG
SDP
Supplier:  Medchemexpress LLC HY14865S100UG
Encompass_Preferred
Omadacycline-d9 is a deuterium-labeled form of Omadacycline, designed for use as an internal standard in analytical processes. It is intended for research purposes only and not for medical applications.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-59952