Learn More
Medchemexpress LLC Omadacycline hydrochloride | 1196800-39-1 | 99.47% | 593.11 | 500 MG
Supplier: Medchemexpress LLC HY14865C500MG
Omadacycline hydrochloride is a first-in-class orally active aminomethylcycline antibacterial, belonging to the tetracycline class of antibiotics. It inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit. It exhibits broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. It is used for research on acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.
- Inhibits bacterial protein synthesis
- Broad-spectrum antibacterial activity
- Active against Gram-positive, Gram-negative, and atypical bacteria
- Effective against MRSA and VRE
- Shows efficacy in mouse systemic infection models