Learn More
Medchemexpress LLC Omadacycline (hydrochloride) | 1196800-39-1 | 99.5% | 593.11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14865C50MG
Encompass_Preferred
Omadacycline hydrochloride is a first-in-class orally active aminomethylcycline antibacterial, part of the tetracycline class of antibiotics. It inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit.
  • Broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria.
  • Used for research on acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.
  • Active against strains expressing tetracycline and other antibiotic resistance.
Catalog No. 50-003-61148