Learn More
Medchemexpress LLC Omega-3 arachidonic acid | 24880-40-8 | MFCD00216033 | 99.0% | 304.47 | C20H32O2 | 500UG
SDP
Supplier:  Medchemexpress LLC HY125140500UG
Encompass_Preferred
ω-3 Arachidonic acid (8Z,11Z,14Z,17Z-eicosatetraenoic acid) is a rare omega-3 polyunsaturated fatty acid used as a research reagent in biochemical and pharmacological studies. It has been reported to inhibit arachidonoyl-CoA synthetase and is supplied in small premeasured vials for in-vitro assays. Typical specifications include 99.0% purity, molecular formula C20H32O2, and molecular weight 304.47 g/mol.
  • Rare omega-3 polyunsaturated fatty acid.
  • Inhibits arachidonoyl-CoA synthetase in biochemical assays.
  • High purity: 99.0%.
  • Supplied as a 500 μg vial in ethanol for convenient dosing.
  • Molecular formula C20H32O2, molecular weight 304.47 g/mol.
Catalog No. 50-002-37649