Learn More
Carbosynth LLC. omega-Conotoxin MVIIC (0.1mg vial), 1ea, Disulfide bonds: (Disulfide bonds between Cys1-Cys16,Cys8-Cys20, and Cys15-Cys26), MW: 2749.3, Molecular formula: C106H178N40O32S7, CAS# 147794-23-8
SDP
Supplier:  Carbosynth LLC. PCN4283S0.1MG
Encompass_Preferred
Synonyms: Cys-Lys-Gly-Lys-Gly-Ala-Pro-Cys-Arg-Lys-Thr-Met-Tyr-Asp-Cys-Cys-Ser-Gly-Ser-Cys-Gly-Arg-Arg-Gly-Lys-Cys-NH2, Storage Temperature: 2-10°C
Catalog No. 50-168-6383