Learn More
Medchemexpress LLC Omeprazole-d3-1 | 934293-92-2 | 95.1% | C17H16D3N3O3S | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0113S11MG
Encompass_Preferred
Omeprazole-d3-1 is a deuterium-labeled form of Omeprazole, a proton pump inhibitor (PPI) used for treating acid-related gastrointestinal disorders. It is also a potent brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor.
  • Competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM.
  • Inhibits the growth of Gram-positive and Gram-negative bacteria.
  • Can be used as a tracer in research.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32381