Learn More
Sigma Aldrich Fine Chemicals Biosciences Omeprazole solid500MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences O104500MG
Encompass
Omeprazole is a benzimidazole derivative a weak base and is lipophilic in nature. This compound is pH sensitive and does not exists in its original form at low pH.
Catalog No. 50-290-2198