Learn More
Medchemexpress LLC Omiganan 25mg | 204248-78-2 | 25MG
SDP
Supplier:  Medchemexpress LLC HY10504825MG
Encompass_Preferred
Omiganan is a cationic antimicrobial peptide Omiganan as an analogue of indolicidin shows activity against gram-positive and gram-negative bacteria but also Candida spp isolates Omiganan can be used for the research of alcohol nose and acne[1 [2
Catalog No. 50-002-44131