Learn More
Enzo Life Sciences OMNIMMP fluorogenic control (1 mg) CAS: 140430-55-3
SDP
Supplier:  Enzo Life Sciences BMLP1270001
Encompass_Preferred
Reference peptide for OMNIMMP fluorogenic substrate (BML-P126) and TACE (ADAM17) fluorogenic substrate (BML-P132). Ex.: 328nm, Em.: 393nm. The following Ex/Em have also been used: 320-340/400-420. Alternative Name: Mca-Pro-Leu-OH. Formula: C23H26N2O7. MW: 444.5. CAS: 140430-55-3. Purity: ≥98% (TLC). Sequence Mca-Pro-Leu-OH [Mca=(7-methoxycoumarin-4-yl)acetyl]. Appearance: White powder. Solubility: Soluble in methanol (50 mg/ml) or DMSO (20 mg/ml). Long Term Storage: -20°C. Use/Stability: Stock solution in methanol is highly stable at -20°C. Handling: Protect from light and moisture. Store under inert gas.
Catalog No. NC1472256