Learn More
Electron Microscopy Sciences Omniprobe Lift-Out Grid 4-Post, Cooper
SDP
Supplier:  Electron Microscopy Sciences 7596410
Encompass_Preferred
4 post copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations, two with vertical bar attachment surfaces and two with "V" shaped alighment surfaces. Sides have lower profile for easier access to outermost posts. 3 mm dia. Packaged in plastic vials.
Material: Copper
Catalog No. 50-190-2654