Learn More
Selleck Chemical LLC ON-013100 E7822-1MG
SDP
Supplier:  Selleck Chemical LLC E78221MG
Encompass_Preferred
ON-013100 is a benzylstyrylsulfone antineoplastic agent and a potent mitotic inhibitor It inhibits CDK4 and cyclin D1 expression while suppressing MDM2 and reducing p53 levels disrupting mitogenic signaling and enhancing its anticancer activity
Catalog No. 50-000-42155