Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED ON-TARGETplus NFE2L2 siRNA Reagents - Human, SMARTPool, 5nmol
Supplier:  HORIZON DISCOVERY BIOSCIENCES LIMITED L003755000005
Encompass
The patented ON-TARGETplus modification pattern for specificity, combined with the SMARTselection algorithm for efficient, guaranteed target gene silencing, makes ON-TARGETplus siRNA the premium choice for optimal knockdown and reduced off-targets.
Catalog No. NC1395340