Learn More
Selleck Chemical LLC Onalespib (AT13387) S1163-5mg
SDP
Supplier:  Selleck Chemical LLC S11635MG
Encompass_Preferred
Onalespib (AT13387) is a selective potent Hsp90 inhibitor with IC50 of 18 nM in A375 cells displays a long duration of anti-tumor activity Phase 2
Catalog No. 50-311-1044