Learn More
Cayman Chemical Onalespib Inhibitors
SDP
Supplier:  Cayman Chemical 106555 MG
Encompass_Preferred
A selective inhibitor of Hsp90 (IC50 = 18 nM); induces the degradation of specific Hsp90 client proteins; efficacious in a range of xenograft models
Catalog No. 50-289-1210