Learn More
Medchemexpress LLC Onartuzumab (Synonyms: MetMAb) 10mg
SDP
Supplier:  Medchemexpress LLC HYP9925010MG
Encompass_Preferred
Onartuzumab (MetMAb) is a unique, humanized and affinity-matured monovalent (one-armed) monoclonal antibody against the MET receptor. Onartuzumab potently inhibits HGF binding and receptor phosphorylation and signaling. Onartuzumab has antibody-like pharmacokinetics and antitumor activity.
Catalog No. NC3397894