Learn More
Medchemexpress LLC HY-111343 10mg Medchemexpress, ONC212 CAS:1807861-48-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11134310MG
Encompass_Preferred
Medchemexpress, HY-111343 10mg ONC212 CAS:1807861-48-8 ONC212, a fluorinated-ONC201 analogue, is a promising anti-cancer agent and also a selective agonist of GPR132. ONC212 also induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8810
 Restricted Item