Learn More
Medchemexpress LLC ONC212 | 1807861-48-8 | 99.86% | 440.46 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111343
Encompass_Preferred
ONC212, a fluorinated-ONC201 analogue, is a promising anti-cancer agent and a selective agonist of GPR132. It also induces apoptosis.
  • Fluorinated-ONC201 analogue.
  • Promising anti-cancer agent.
  • Selective agonist of GPR132.
  • Induces apoptosis.
Catalog No. 50-003-47000