Learn More
Medchemexpress LLC Oncrasin-72 | 92407-90-4 | 97.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130207100MG
Encompass_Preferred
Oncrasin-72 is an RNA polymerase II inhibitor that inhibits growth and induces cell death in human cancer cells. It demonstrates antitumor activity through JNK activation and STAT3 inhibition. A validated analytical method for this compound is used to quantify its concentration in biological fluids for pharmacokinetic studies, and this method successfully quantified it in rat plasma across different dose groups.
  • RNA polymerase II inhibitor
  • Inhibits growth and induces cell death in human cancer cells
  • Exhibits antitumor activity through JNK activation and STAT3 inhibition
  • Validated analytical method for pharmacokinetic studies
  • For research use only
Catalog No. 50-003-39925