Learn More
TARGETMOL CHEMICALS INC Ondansetron 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6615200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Ondansetron (GR 38032) is a serotonin 5-HT3 receptor antagonist used mainly as an antiemetic. Purity 98.9%
Catalog No. 50-287-7103