Learn More
Enzo Life Sciences Ondansetron (50mg). CAS: 99614-02-5
SDP
Supplier:  Enzo Life Sciences BMLNS1450050
Encompass_Preferred
A serotonin 5-HT3 receptor antagonist. Clinically effective anti-emetic especially in chemotherapy-induced nausea. Purity: ≥98% (NMR). Solubility: Soluble in DMSO (10 mg/ml warm). Long Term Storage: Ambient.
Catalog No. 50-201-1214