Learn More
New England Biolabs, Inc. OneTaq Hot Start 2X Master Mix with GC Buffer – 100 reactions (50 µl vol)
SDP
Supplier:  New England Biolabs, Inc. M0485S/DEL
Encompass
OneTaq Hot Start 2X Master Mix with GC Buffer is an optimized blend of Taq and Deep Vent DNA Polymerases combined with an aptamer-based inhibitor. This enzyme blend is ideally suited to PCR applications from GC-rich templates, including pure DNA solutions, bacterial colonies and cDNA products. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient master mix formulation contains dNTPs, MgSO4, buffer components and stabilizers, requiring only the addition of primers and DNA template for robust amplification.
Catalog No. NC0108403