Learn More
Medchemexpress LLC ONO-5334 5mg | 868273-90-9 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1080445MG
Encompass_Preferred
ONO-5334 is a potent selective and orally active cathepsin K inhibitor with Ki values of 0 10 nM 0 049 nM and 0 85 nM for human rabbit and rat cathepsin K respectively ONO 5334 is an effective antiviral compound against SAR-COV-2 virus activity with an EC50 value of 500 nM ONO-5334 has the potential for the study of osteoporosis and COVID-19 disease[1
Catalog No. 50-002-41909