Learn More
Medchemexpress LLC Ono-AE3-208 | 402473-54-5 | MFCD13184814 | >98.3% | C24H21FN2O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY5090110MM/1ML
Encompass_Preferred
ONO-AE3-208, also known as AE 3-208, is a selective and orally active EP4 receptor antagonist with a Ki of 1.3 nM. It demonstrates less potent effects on EP3, FP, and TP receptors. This compound is capable of suppressing cell invasion, migration, and metastasis of prostate cancer, showing dose-dependent inhibition of cell invasion and migration in vitro without affecting proliferation. In vivo studies indicate that ONO-AE3-208 suppresses the bone metastasis of PC3 cells in mice.
  • Suppresses cell invasion, migration, and metastasis of prostate cancer
  • Inhibits cell invasion and migration in a dose-dependent manner in vitro
  • Does not affect cell proliferation in vitro
  • Abolishes CTGF in the presence of the EET synthesis inhibitor MS-PPOH
  • Blocks arachidonic acid (AA)-induced dilation of the attached Af-Art
  • Suppresses bone metastasis of PC3 cells in mice
Catalog No. 50-003-57748